Learn More
Magnesium Ethylate, Sigma-Aldrich™
Product Code. 15870671
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870671 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15870671 Supplier Sigma Aldrich Supplier No. 8188940100
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2414-98-4
Specifications
MDL Number MFCD00009060
Quantity 100 g
Synonym Magnesium ethylate, Magnesium ethylate
Molecular Weight (g/mol) 114.43
Grade Research