missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acrylate

Product Code. p-7051801
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305017 5 g 5g
11395007 25 g 25g
11385007 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11305017 Supplier Thermo Scientific Alfa Aesar Supplier No. 042002.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5698-98-6 | C6H6MgO4 | 166.415 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5698-98-6
Molecular Formula C6H6MgO4
Molecular Weight (g/mol) 166.415
MDL Number MFCD00068285
InChI Key DWLAVVBOGOXHNH-UHFFFAOYSA-L
Synonym magnesium acrylate, unii-38v94j49hw, 2-propenoic acid, magnesium salt 2:1, 2-propenoic acid, magnesium salt, 79-10-7 parent, magnesium di-acrylate, magnesium acrylate 5g, acmc-1alvh, magnesium prop-2-enoate, diacrylic acid magnesium salt
PubChem CID 165336
IUPAC Name magnesium;prop-2-enoate
SMILES C=CC(=O)[O-].C=CC(=O)[O-].[Mg+2]

Specifications

Melting Point >220°C (decomposition)
Linear Formula [H2C=CHCO2]2Mg
Quantity 5 g
Sensitivity Air and moisture sensitive
Formula Weight 166.42
Physical Form Powder
Chemical Name or Material Magnesium acrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.