Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Wash Solution
Product Code. 16305035
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16305035 1000 mL 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16305035 Supplier Applied Biosystems™ Supplier No. A42360
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
This wash solution is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
This wash solution is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more wash solution than is provided in the kit.
Important: This wash solution may develop inert white or brown particulates that float in solution. This is not a cause for concern and does not negatively affect performance.
TRUSTED_SUSTAINABILITY
Specifications
Sample Type Serum, Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Capacity 1000 mL
Content And Storage Store at room temperature.
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Purification Target Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids
Quantity 1000 mL
Shipping Condition Room Temperature