Learn More
Applied Biosystems™ MagMAX™ Microbiome Bead Tubes
Description
These bead tubes are included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42358). They are made available separately for applications that require more tubes than are provided in the kit.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Sample Type | Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| Product Line | MagMAX |
| Quantity | 100 tubes |
| Shipping Condition | Room Temperature |
| Target | Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids |