Learn More
Thermo Scientific™ MagJET Genomic DNA Kit
Product Code. 13575170 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
96 Preps
Unit Size:
96 tests
Product Code. No. of Reactions unitSize
13575170 96 Preps 96 tests
1 options
Product Code. 13575170
Brand: Thermo Scientific™ K2721
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Purify genomic DNA from cell culture, tissue samples, bacteria, and yeast with a magnetic bead separation method amenable to high-throughput workflows.
MagJET Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from tissue and cell cultures, bacteria and yeast, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles.
The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users.
The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors, can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.
Highlights
  • Versatile -suitable for gDNA purification from variety of biological samples, including gDNA from eukaryotic cells, animal tissues, yeast and bacteria, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles
  • Efficient -high yields of high molecular weight genomic DNA
  • High quality -isolates high integrity gDNA, free of inhibitors and RNA contamination
  • Flexible -easily adjustable for high throughput automation platforms and manual processing
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Ambient
No. of Reactions 96 Preps