Learn More
Invitrogen™ MagicMedia™ E. coli Expression Medium, Powder
Product Code. p-7180343
Click to view available options
Quantity:
1 L
5 x 1 L
Unit Size:
1 set
1kg
Product Code. 15351532
Brand: Invitrogen™ K6810
Includes
Contains a proprietary Component A (in foil pouch powder format), and Component B (in buffer liquid format).
MagicMedia E. coli Expression Medium is specifically designed to dramatically increase the yield of recombinant proteins in T7-regulated E. coli expression systems without monitoring culture OD (optical density) or adding inducer. The proprietary MagicMedia formulation enables E. coli growth to reach culture densities 3- to 10-fold higher than those achieved with traditional LB + IPTG methods, allowing you to:
• Achieve 3–10 times higher protein yields (Figure 1) with much less hands-on time
• Eliminate time-consuming OD monitoring steps—simply inoculate, grow the culture overnight, and harvest (Figure 2)
• Reduce culture volume to simplify handling and scale-up processes
• Ideal for simultaneous expression of multiple clones for high-throughput applications
MagicMedia E. coli Expression Medium, powder, consists of a proprietary Component A (in foil pouch powder format) and Component B (in buffer liquid format).
MagicMedia E. coli Expression Medium is also available in a ready-to-use, pre-sterilized liquid format (Cat. No. K6803).
Order Info
Store Component A (foil pouches) and Component B buffer (liquid) at Room Temperature (15 - 30°C).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store Component A (foil pouches) and Component B (liquid) at room temperature (15–30°C).
Form Powder, Liquid
Format Kit
Product Type Microbiology Media
Product Line MagicMedia
Quantity 1 L
Species Escherichia coli