Learn More
MAGEB4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12786-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis.Specifications
| MAGEB4 | |
| Polyclonal | |
| Unconjugated | |
| MAGEB4 | |
| CT3.6, MAGE B4 antigen, MAGEB4, melanoma antigen family B, 4, Melanoma associated antigen B4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4115 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O15481 | |
| MAGEB4 | |
| MAGEB4 Fusion Protein Ag3502 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |