Learn More
Abnova™ MAGEA1 (Human) Recombinant Protein
Product Code. p-7164439
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16127551
Brand: Abnova™ H00004100P01.10ug
Human MAGEA1 full-length ORF ( AAH17555, 1 a.a. - 309 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MSLEQRSLHCKPEEALEAQQEALGLVCVQAAASSSSPLVLGTLEEVPTAGSTDPPQSPQGASAFPTTINFTRQRQPSEGSSSREEEGPSTSCILESLFRAVITKKVADLVGFLLLKYRAREPVTKAEMLESVIKNYKHCFPEIFGKASESLQLVFGIDVKEADPTGHSYVLVTCLGLSYDGLLGDNQIMPKTGFLIIVLVMIAMEGGHAPEEEIWEELSVMEVYDGREHSAYGEPRKLLTQDLVQEKYLEYRQVPDSDPARYEFLWGPRALAETSYVKVLEYVIKVSARVRFFFPSLREAALREEEEGV
Specifications
Accession Number AAH17555
Gene ID (Entrez) 4100
Name melanoma antigen family A, 1 (directs expression of antigen MZ2-E)
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias MAGE1, MGC9326
Gene Symbol MAGEA1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer