Learn More
MP BIOMEDICALS MagBeads FastDNA/RNA Kit for Pathogen 50 Preps
Shop All MP Biomedicals Products
Click to view available options
Isolation Technology:
Magnetic Bead
Unit Size:
1 set
Description
- The MagBeads FastDNA/RNA Kit for Pathogen offers an advanced rapid nucleic acid extraction solution based on next-generation silicon-based magnetic bead technology
- Delivers high-fidelity gDNA/RNA extraction from samples such as bodily fluids for detection requirements
- Features a streamlined extraction protocol requiring only a single liquid-transfer step prior to automation
- Includes an optimized lysis protocol ensuring complete and consistent bacterial cell disruption for reliable nucleic acid detection
- Effectively prevents exogenous nucleic acid contamination and ensures accurate sample composition
- Provides an accurate reflection of the sample's intrinsic species composition
- Produces high-purity nucleic acid suitable for a wide array of downstream molecular applications
- Well-suited for PCR
- Well-suited for RT-PCR
- Well-suited for construction of metagenomic libraries
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| For Use With (Application) | DNA/RNA Purification |
| Product Type | Nucleic Acid Extraction Kit |