missing translation for 'onlineSavingsMsg'
Learn More

Magainin 1, MedChemExpress

Product Code. 16479428
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. Quantity unitSize
16479428 10 mg 10mg
16491918 5 mg 5mg
16412739 1 mg 1mg
3 options
This item is not returnable. View return policy

Product Code. 16479428

Brand: Medchem Express HYP0269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Magainin 1 (Magainin I) is an antimicrobial and amphipathic peptide isolated from the skin of Xenopus laevis. Magainin 1 exhibits antibiotic activity against numerous Gram-negative and Gram-positive bacteria.

Magainin 1 (Magainin I) is an antimicrobial and amphipathic peptide isolated from the skin of Xenopus laevis. Magainin 1 exhibits antibiotic activity against numerous Gram-negative and Gram-positive bacteria.

Specifications

Chemical Name or Material Magainin 1
Physical Form Powder
CAS 108433-99-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C112H177N29O28S
For Use With (Application) COVID-19-immunoregulation
Synonym Magainin I
SMILES [GIGKFLHSAGKFGKAFVGEIMKS]
Molecular Weight (g/mol) 2409.85
Formula Weight 2409.85
Percent Purity 99.66%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.