Learn More
Mafenide Hydrochloride, LoGiCal
Product Code. 30458221
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458221 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458221 Supplier LoGiCal Supplier No. LGCFOR3127.00
 Restricted Item
Estimated Shipment: 30-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Especificaciones
Chemical Name or Material Mafenide Hydrochloride
CAS 138-37-4
Analyte or Component Names Mafenide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C7 H10 N2 O2 S . Cl H
Certifications/Compliance ISO 9001
Synonym Benzenesulfonamide, 4-(aminomethyl)-, hydrochloride (1:1), Benzenesulfonamide, 4-(aminomethyl)-, monohydrochloride (9CI), 4-(Aminomethyl)benzene sulfonamide monohydrochloride, 4-Aminosulfonylbenzylamine hydrochloride, 4-Homosulfanilamide hydrochloride, Homosulfamine, Homosulfamine hydrochloride, Homosulfanilamide hydrochloride, Mafenide hydrochloride, Marfanil, Sulfamylon hydrochloride, p-(Aminomethyl)benzenesulfonamide hydrochloride, p-(Aminomethyl)benzenesulfonamide monohydrochloride, p-Aminomethylphenylsulfonamide hydrochloride, p-Toluenesulfonamide, alpha-amino-, hydrochloride (6CI), p-Toluenesulfonamide, alpha-amino-, monohydrochloride (8CI), alpha-Amino-p-toluenesulfonamide hydrochloride
InChI Formula InChI=1S/C7H10N2O2S.ClH/c8-5-6-1-3-7(4-2-6)12(9,10)11;/h1-4H,5,8H2,(H2,9,10,11);1H
Solution Type Neat
SMILES Cl.NCc1ccc(cc1)S(=O)(=O)N
IUPAC Name 4-(aminomethyl)benzenesulfonamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 222.69
Quantity 10 mg
Formula Weight 222.023 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.