Learn More
Cytiva Macrosep™ Advance Centrifugal Devices with Omega Membrane
Product Code. p-7089754
Change view
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
3 kDa
30 kDa
Includes:
100/Pk.
24/Pk.
Color:
Blue
Clear
Gray
Red
Yellow
Unit Size:
Pack of 100
Pack of 24
Product Code. MWCO Includes Color unitSize
17154731 3 kDa 100/Pk. Gray Pack of 100
17174731 10 kDa 100/Pk. Blue Pack of 100
17124731 1 kDa 24/Pk. Yellow Pack of 24
17184731 30 kDa 24/Pk. Red Pack of 24
17104741 100 kDa 24/Pk. Clear Pack of 24
17134731 1 kDa 100/Pk. Yellow Pack of 100
17164731 10 kDa 24/Pk. Blue Pack of 24
17114741 100 kDa 100/Pk. Clear Pack of 100
17194731 30 kDa 100/Pk. Red Pack of 100
17144731 3 kDa 24/Pk. Gray Pack of 24
10 options
Product Code. 17154731
Brand: Cytiva MAP003C38
Only null left
Add to basket
Add to basket
Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.
  • Exchange buffer or remove salt of chromatography eluates and gradient fractions
  • For research applications only
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Description EFA: 7.2 cm2
Material (Membrane) PES
MWCO 3 kDa
Sterility Non-sterile
Includes 100/Pk.
Volume (Metric) 5 to 20 mL (Sample)