Learn More
Berthold Technologies Macro Lens 50 mm F0.95 Ikon Version
Product Code. 31777437 Shop All Berthold Technologies Products
Change view
Click to view available options
Width (Metric):
50 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
31777437 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31777437 Supplier Berthold Technologies Supplier No. 60223
 Restricted Item
Estimated Shipment: 20-11-2026
Used for closeup imaging in the NightSHADE in vivo imaging system.
  • Required when using the Macro table with the NightSHADE system
  • Enables image acquisition at higher magnification
Specifications
Type Macro Lens
For Use With (Equipment) Imaging System
Width (Metric) 50 mm