Learn More
MA-0204, MedChemExpress
Product Code. 16473256
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16473256
Brand: Medchem Express HY114739
MedChemExpress MA-0204 is a potent, highly selective and orally available peroxisome proliferator activated receptor δ (PPARδ) modulator with EC50s of 0.4 nM, 7.9 nM and 10 nM for human, mouse and rat PPARδ, respectively. Potential treatment for Duchene Muscular Dystrophy (DMD).
MA-0204 is a potent, highly selective and orally available peroxisome proliferator activated receptor δ (PPARδ) modulator with EC50s of 0.4 nM, 7.9 nM and 10 nM for human, mouse and rat PPARδ, respectively. Potential treatment for Duchene Muscular Dystrophy (DMD).
Specifications
Chemical Name or Material MA-0204
CAS 2095128-17-7
Color White
Purity Grade Notes Research
Molecular Formula C25H27F3N2O4
Quantity 25 mg
Solubility Information DMSO : 43.33 mg/mL (90.94 mM; Need ultrasonic)
SMILES O=C(C[C@@H](CCCOC1=CC=CC=C1CN2C(C)=CN=C2C3=CC=C(C=C3)OC(F)(F)F)C)O
Molecular Weight (g/mol) 476.49
Formula Weight 476.49
Percent Purity 99.31%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.