Learn More
Brady™ M710 Label Printer with BWS SFID Suite
Product Code. 17845255
Click to view available options
Keyboard Type:
AZERTY
QWERTY (EU)
Unit Size:
Each
Product Code. 17845255
Brand: Brady™ 317826
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
More speed, less mess. It’s Brady™'s fastest, most advanced portable printer yet with a print speed of 76.2 mm/sec. Printer includes Brady™ WorkStation Product & Wire Identification Suite.
  • Auto-cut labels and work all day on one charge
  • Use one device for 100's of applications, printing on the toughest materials around
  • Get lightning-fast print speeds, plus cut labels automatically
  • Work longer – print 4,500 labels on a full battery charge
  • Enjoy more durability with a printer that withstands 1.2 m drops, blowing sand and dust
  • Print what you need, how you need – text, barcodes, shapes and images using a keypad or software
  • Get shop-to-field flexibility with label saving, sharing and data imports
  • Comes with a Brady™ WorkStation Safety & Facility Identification Suite, enabling you to immediately start designing professional labels
Specifications
Product Type Printer
Color Blue
Display Type Color LCD Touchscreen
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Length (English) 4.6 in.
Width (English) 9.45 in.
Length (Metric) 116.75 mm
Width (Metric) 240.03 mm
Height (English) 13.59 in.
Height (Metric) 345.2 mm
Color Capability Monocolor
Display Size 53.77 x 71.28 mm
Includes Printer, Hard Case, USB Cable, Quick Start Guide, Drivers, Li-ion Rechargeable Battery, AC Adapter, Users Guide, Brady WorkStation Safety and Facility ID Software Suite Download, Media Adapter, Roll of M7C-1000-595-WT and M7-R6000 Ribbon
Languages English (US), French, Spanish, Portuguese, German, Dutch, Norwegian, Finnish, Swedish, Italian, Danish, Bulgarian, Russian, Croatian, Hungarian, Japanese, Korean, Polish, Romanian, Slovak, Turkish, Estonian, Chinese (Traditional), Chinese (Simplified)
Memory 200 MB (Stores Up to 100,000 Labels)
Model M710
Print Speed 76.2 mm/sec.
Resolution 300 dpi
Connector Types USB 2.0, USB-C
Weight (English) 5.4 lb.
Weight (Metric) 2.45 kg
Certifications/Compliance FCC Part 15 Class A
For Use With (Application) Facility Identification, Safety Identification
Warranty 2 Years
Power Supply Details AC Adaptor, Rechargeable Battery
Voltage 110/240 V
Keyboard Type AZERTY
Alternate Part Number M710-AZ-SFID