Learn More
m-Tolylhydrazine, 97%, Thermo Scientific™
Product Code. 11338525
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338525 10 g 10g
11328525 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11338525 Supplier Thermo Scientific Alfa Aesar Supplier No. L11958.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
m-Tolylhydrazine is used as pharmaceutical intermediate. It reacts with cyclohexanone and obtain the 7-methyl-1,2,3,4-tetrahydro-carbazole.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 536-89-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00047815
InChI Key GPTOGZLZMLJZCV-UHFFFAOYSA-N
Synonym m-tolylhydrazine, 3-methylphenyl hydrazine, hydrazine, 3-methylphenyl, m-tolylhydrazin, m-toluidine amine, m-tolyl hydrazine, m-tolyl-hydrazine, 1-m-tolylhydrazine, 3-methylphenylhydrazinehydrochloride, acmc-20akdk
PubChem CID 68308
IUPAC Name (3-methylphenyl)hydrazine
SMILES CC1=CC=CC(NN)=C1
Specifications
Density 1.057
Boiling Point 132°C to 134°C (16 mmHg)
Refractive Index 1.589
Quantity 10 g
UN Number UN2810
Beilstein 742264
Merck Index 14,9542
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material m-Tolylhydrazine