Learn More
Brady™ M Series Indoor/Outdoor Industrial Vinyl Label Supply
Click to view available options
Color:
Black on Clear
White on Clear
Dimensions (L x W):
20 ft. x 1 in.
25 ft. x 1 in.
Unit Size:
Each
Description
- Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces--including outdoors
- Conforms and adheres to difficult surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
- Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
- All-in-one cartridge includes ribbon and label supply
Specifications
Specifications
| Product Type | Label |
| Color | Black on Clear |
| For Use With (Equipment) | BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer |
| Material | Vinyl |
| Print Technology | Thermal Transfer |
| Length (English) | 20 ft. |
| Width (Metric) | 68.58 mm |
| Dimensions (L x W) | 20 ft. x 1 in. |
| Adhesive Type | Permanent Acrylic |
| Autoclavable | Not Autoclavable |
| Show More |