Learn More
Labconco™ Lyph-Lock™ Flask Top
Product Code. p-7111363
Click to view available options
Capacity (Metric):
25 mL, 50 mL
100 mL, 250 mL
500 mL, 750 mL, 1000 mL
Standard Taper Joint:
19/38
20/40
Unit Size:
Each
Product Code. 15143045
Brand: Labconco™ 7556000
Only null left
Add to basket
Add to basket
Three-piece flasks for freeze drying samples. Provides a high-vacuum seal for efficient lyophilization.
Select your Lyph-Lock Flasks based on your sample sizes. Flasks should be filled to no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is ensured.
  • Flexible, silicone rubber seal grasps top and bottom and holds them securely together during pre-freezing and lyophilization
  • A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring seal
  • Top and bottom are high-strength borosilicate glass
  • Seal is a flexibile, non-contaminating silicone rubber
  • Tops, bottoms and seals are available separately as replacement components
  • Adapters are required to attach flasks to freeze dry valve ports (sold separately)
  • Borosilicate glass has maximum temperature is 490° C and silicone maximum temperature is 450° C
  • Flat bottom reduces spills and provides better balance
  • Compatible with all major brands of laboratory freeze dryers
  • Entire flask may be autoclaved
Specifications
Capacity (Metric) 25 mL, 50 mL
Material Borosilicate Glass
For Use With (Equipment) 25 and 50mL Lyph-Lock Flask Bottom and Seal
Standard Taper Joint 20/40