missing translation for 'onlineSavingsMsg'
Learn More

LY3009120, MedChemExpress

Product Code. p-200095843
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16491457 50 mg 50mg
16485248 10 mg 10mg
16477967 5 mg 5mg
16495916 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16491457

Brand: Medchem Express HY1255850mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY3009120 (DP-4978) is a pan RAF inhibitor which inhibits BRAFV600E, BRAFWT and CRAFWT with IC50s of 5.8, 9.1 and 15 nM, respectively.

LY3009120 (DP-4978) is a pan RAF inhibitor which inhibits BRAFV600E, BRAFWT and CRAFWT with IC50s of 5.8, 9.1 and 15 nM, respectively.

Specifications

Chemical Name or Material LY3009120
CAS 1454682-72-4
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C23H29FN6O
Quantity 50 mg
Synonym DP-4978
Solubility Information DMSO : ≥ 38 mg/mL (89.51 mM)
SMILES O=C(NC1=CC(C2=CC3=CN=C(NC)N=C3N=C2C)=C(C)C=C1F)NCCC(C)(C)C
Molecular Weight (g/mol) 424.51
Formula Weight 424.51
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.