Learn More
MedChemExpress LY2334737
Specifications
Specifications
| Molecular Weight (g/mol) | 389.39 g/mol |
| Product Type | Reference compound |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 99.02% |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (256.81 mM) |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 892128-60-8 |