Learn More
Invitrogen™ Ly-6G Monoclonal Antibody (1A8-Ly6g), Biotin, eBioscience™, Invitrogen™

Description
Description: The monoclonal antibody 1A8-Ly6g reacts with mouse Ly-6G, which is also known as Gr-1. Ly-6G is a 25-kDa GPI protein expressed exclusively by neutrophils. Studies suggest an important role in neutrophil infiltration, recruitment and migration. Ly-6G associates with B2 integrins CD11a and CD11b and can attenuate B2 integrin expression and function. The ligand for Ly-6G is currently unknown. Unlike the RB6-8C5 antibody, 1A8-Ly6g is reported to only bind to Ly-6G found on neutrophils. Moreover, 1A8-Ly6g has been used to deplete neutrophils and reported to have antagonist activity. Applications Reported: This 1A8-Ly6G antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1A8-Ly6G antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Specifications
Specifications
| Antigen | Ly-6G |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1A8-Ly6g |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Ly6g |
| Gene Accession No. | P35461 |
| Gene Alias | Gr1; Gr-1; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.