missing translation for 'onlineSavingsMsg'
Learn More

LX2343, MedChemExpress

Product Code. 16407167
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16407167

Brand: Medchem Express HY111383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LX2343 is a BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM. LX2343 acts as a non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM. LX2343 stimulates autophagy in its promotion of Aβ clearance.

LX2343 is a BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM. LX2343 acts as a non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM. LX2343 stimulates autophagy in its promotion of Aβ clearance.

Specifications

Chemical Name or Material LX2343
CAS 333745-53-2
Color White
Purity Grade Notes Research
Molecular Formula C22H19ClN2O6S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (210.57 mM)
SMILES O=C(NC1=CC=C(OCO2)C2=C1)CN(C3=CC(Cl)=CC=C3OC)S(=O)(C4=CC=CC=C4)=O
Molecular Weight (g/mol) 474.91
Formula Weight 474.91
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.