Learn More
Motic™ Camera Adapter, C-Mount, 1X, 1in.
Choose the most appropriate adapter for your camera and microscope and unlock the potential of photomicography. Motic™ Camera Adapter, C-Mount, 1X, 1in.uses the industry standard attachment for digital imaging devices dedicated to microscopes.
Brand: Motic™ 1101001901771
Additional Details : Weight : 0.80000kg
Description
- Connect a 1-inch sensor C-mount camera with this adapter.
- Suitable for BA Series Upright, AE Series Inverted and SMZ Series Stereo Microscopes
Specifications
C-Mount Camera Adapter | |
C-Mount | |
BA Series Upright, AE Series Inverted, SMZ Series Stereo Microscopes, 1in. Sensor C-Mount Camera | |
1X |