Learn More
Welch™ LVS 302 Z Laboratory Vacuum System
Product Code. 11842253 Shop All Welch Ilmvac Products
Change view
Click to view available options
Depth (Metric):
310 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) unitSize
11842253 310 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11842253 Supplier Welch™ Supplier No. 115043
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Electrical Requirements 230 V 50/60 Hz
Depth (Metric) 310 mm
Model LVS 302 Z
Frequency 50/60 Hz
Overall Height (Metric) 445 mm
Overall Width (Metric) 360 mm
Pump Type Laboratory Vacuum System
Maximum Pressure <8 mbar
Voltage 230 V