Learn More
Novus Biologicals™ Lung Tissue Slides (Normal)- Paraffin
Product Code. 18718193 Shop All Bio Techne Products
Change view
Click to view available options
Lung Tissue Slides (Normal)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18718193 Lung Tissue Slides (Normal)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18718193 Supplier Novus Biologicals™ Supplier No. NBP230182
  • ONLINE EXCLUSIVE PRICE
    2740.00 NOK
    2591.40 NOK / Pack of 5
    Save 148.60 NOK
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-10-2026
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Applications: Immunohistochemistry-Paraffin
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
TRUSTED_SUSTAINABILITY
Specifications
Cross Reactivity Human
Type Lung Tissue Slides (Normal)
For Use With (Application) Immunohistochemistry-Paraffin
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Normal