Learn More
Merck Immobilon Western HRP Chemiluminescence Substrates
Product Code. 11790644 Shop All Merck Products
Click to view available options
Quantity:
500 mL
Product Type:
HRP Substrate
Sensitivity:
∼1 to 3 pg
∼400 fg
∼6 pg
Unit Size:
500mL
Each
Product Code. 11790644
Brand: Merck WBLUR0500
Premixed, ready-to-use chemiluminescent reagents for detection of HRP-based Westerns
Horseradish peroxidase (HRP) catalyzes the oxidation of luminol in the presence of peroxide, releasing photons (chemiluminescence) that are captured on x-ray films or by CCD cameras. Most HRP substrates are plagued by quickly decaying signal and short shelf life, but Luminata substrates are formulated differently to include stabilizing reagents.
  • Consistent results with less pipetting error
The stabilizers added to Luminata substrates:
  • Prevent spontaneous oxidation of luminol by peroxide in the absence of HRP
  • Enhance the stability of both the luminol and peroxide, ensuring that Luminata substrates are stable for one year in their premixed format
  • Stabilize, strengthen, and prolong the chemiluminescence signal once HRP starts to catalyze oxidation
Three different sensitivities for a range of detection needs:
  • Immobilon Classico premixed reagent for everyday detection needs
  • Immobilon Crescendo premixed reagent for detection in the picogram range
  • Immobilon Forte premixed reagent for detection in the femtogram range
TRUSTED_SUSTAINABILITY
Specifications
Product Type HRP Substrate
For Use With (Application) Dot/Slot Blot
Quantity 500 mL
Sensitivity ∼1 to 3 pg
Content And Storage 2°C to 8°C