Learn More
Merck Luminata Crescendo ELISA HRP Substrate
Used on immunoblots as chemiluminescent substrate
Brand:  Merck ELLUR0200
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11357630
  • 6280.00 NOK / 200mL
Estimated Shipment: 21-05-2024
Description
  • Detection Methods: Chemiluminescence
  • Key Applications: ELISA
  • Chemiluminescent: HRP detection reagent
  • For use on immunoblots
Specifications
Specifications
200 mL
2°C to 8°C