Learn More
Euro Protection™ Lumbar Belt without Straps
Product Code. 12063596
Change view
Click to view available options
Length (Metric):
82 cm
102 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12063596 102 cm Each
12053596 82 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12063596 Supplier Euro Protection™ Supplier No. 78112
  • ONLINE EXCLUSIVE PRICE
    666.48 NOK
    358.06 NOK / Pack of 1
    Save 308.42 NOK
    46% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Euro Protection™ Lumbar Belt without Straps uses variable pressure to provide lumbar support.
  • Variable pressure and fastening system
  • To be worn outside another garment
Specifications
Product Type Lumbar Support Belt
Length (Metric) 102 cm