missing translation for 'onlineSavingsMsg'
Learn More

Lucifer yellow CH, Dilithium salt, ≥99%, >95%, MP Biomedicals

Product Code. 11467532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
Product Code. Quantity unitSize
11467532 10 mg 10mg
11477532 25 mg 25mg
11487532 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 11467532

Brand: MP Biomedicals 0215526710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67769-47-5
Molecular Formula C13H9Li2N5O9S2
Molecular Weight (g/mol) 457.241
InChI Key RPKCZJYDUKVMGF-UHFFFAOYSA-L
Synonym 6-amino-2[(Hyrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-Benz[de] Isoquinoline-5,8-disulfonic acid dilithium salt
PubChem CID 124220108
ChEBI CHEBI:52106
IUPAC Name dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
SMILES [Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]

Specifications

Assay Percent Range >95%
Formula Weight 457.2
Percent Purity ≥99%
Color Orange to Yellow
Quantity 10 mg
Physical Form Powder
Chemical Name or Material Lucifer yellow CH, Dilithium salt, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.