Learn More
Lubiprostone-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Lubiprostone-d7 |
| CAS | 1217675-13-2 |
| Molecular Formula | C20H25D7F2O5 |
| Quantity | 1 mg |
| Synonym | (11α)-16,16-Difluoro-11-hydroxy-9,15-dioxo-prostan-1-oic Acid-d7, Ru 0211-d7 |
| InChI Formula | InChI=1S/C20H32F2O5/c1-2-3-12-20(21,22)18(25)11-10-15-14(16(23)13-17(15)24)8-6-4-5-7-9-19(26)27/h14-15,17,24H,2-13H2,1H3,(H,26,27)/t14-,15-,17-/m1/s1/i1D3,2D2,3D2 |
| SMILES | O=C(C(F)(F)CC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])CC[C@@H]1[C@@H](CCCCCCC(O)=O)C(C[C@H]1O)=O |
| Molecular Weight (g/mol) | 397.51 |