Learn More
IKA™ LT 5.28 Pressure Control Valve
Product Code. 17503626 Shop All IKA Products
Change view
Click to view available options
Pressure Control Valve; For Use With: Temperature Control Jacket of the Laboratory Reactor Vessel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17503626 Pressure Control Valve; For Use With: Temperature Control Jacket of the Laboratory Reactor Vessel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17503626 Supplier IKA™ Supplier No. 0020100227
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
The pressure control valve LT 5.28 is used to regulate the pressure in the temperature control jacket of the laboratory reactor vessel.
  • The pressure control valve LT 5.28 is used to regulate the pressure in the temperature control jacket of the laboratory reactor vessel after the temperature control hoses have been disconnected.
  • The valve prevents the formation of an overpressure, which could result from a temperature change of the medium in the temperature control jacket.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Temperature Control Jacket of The Laboratory Reactor Vessel