missing translation for 'onlineSavingsMsg'
Learn More

LSZ-102, MedChemExpress

Product Code. p-200097064
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16474468

Brand: Medchem Express HY111486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LSZ-102 is a potent, orally bioavailable selective estrogen receptor degrader with an IC50 of 0.2 nM.

LSZ-102 is a potent, orally bioavailable selective estrogen receptor degrader with an IC50 of 0.2 nM.

Specifications

Chemical Name or Material LSZ-102
CAS 2135600-76-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H17F3O4S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (212.56 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)/C=C/C1=CC=C(OC2=C(C3=CC=C(F)C=C3C(F)(F)C)SC4=CC(O)=CC=C42)C=C1
Molecular Weight (g/mol) 470.46
Formula Weight 470.46
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.