missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10700-1-AP-20UL
This item is not returnable.
View return policy
Description
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.Specifications
| LSM5 | |
| Polyclonal | |
| Unconjugated | |
| LSM5 | |
| LSM5, YER146W | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23658, 66373 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62322, Q9Y4Y9 | |
| Lsm5 | |
| LSM5 Fusion Protein Ag1145 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction