missing translation for 'onlineSavingsMsg'
Learn More

LSG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16897704 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16897704 20 μL 20µL
16887704 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16897704

Brand: Proteintech 177501AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein related to the yeast large subunit GTPase 1. The encoded protein is necessary for cell viability and may localize in the endoplasmic reticulum, nucleus and cytoplasm.

Specifications

Antigen LSG1
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LSG1
Gene Accession No. Q3UM18, Q5BJT6, Q9H089
Gene Alias hLsg1, Large subunit GTPase 1 homolog, LSG1
Gene Symbols Lsg1
Host Species Rabbit
Immunogen LSG1 Fusion Protein Ag12016
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224092, 288029, 55341
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.