missing translation for 'onlineSavingsMsg'
Learn More

LRWD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19546-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16849084

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody recognizes the N-term of LRWD1.

Required for G1/S transition. Recruits and stabilizes the origin recognition complex (ORC) onto chromatin during G1 to establish pre-replication complex (preRC) and to heterochromatic sites in post-replicated cells. Binds a combination of DNA and histone methylation repressive marks on heterochromatin. Binds histone H3 and H4 trimethylation marks H3K9me3, H3K27me3 and H4K20me3 in a cooperative manner with DNA methylation. Required for silencing of major satellite repeats. May be important ORC2, ORC3 and ORC4 stability.
Specifications

Specifications

LRWD1
Polyclonal
Unconjugated
LRWD1
LRWD1
Rabbit
Antigen Affinity Chromatography
RUO
222229, 71735
-20°C
Liquid
Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BUI3, Q9UFC0
Lrwd1
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers