missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC52 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25584-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This polyclonal antibody raised against 24-244aa of human LRRC52 reveals a band of 45 kDa, which is consistent with the molecular weight of glycosylated form of LRRC52.
LRRC52 (leucine-rich repeat-containing protein 52) belongs to the gamma family of the BK channel accessory subunits, which potentially regulate the channel’s gating properties. It is domitly expressed in testis and skeletal muscle, with much lower expression in several other tissues including placenta, kidney, lung, and some glands. LRRC52 has been reported as a Slo3 auxiliary subunit that helps define the specific alkalization dependence of KSper activation. LRRC52 has a calculated molecular weight of 35 kDa.Specifications
LRRC52 | |
Polyclonal | |
Unconjugated | |
LRRC52 | |
LRRC52 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
240899, 440699 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5M8M9, Q8N7C0 | |
LRRC52 | |
LRRC52 Fusion Protein Ag22354 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |