missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC32 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26021-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.Specifications
LRRC32 | |
Polyclonal | |
Unconjugated | |
LRRC32 | |
D11S833E, GARP, Garpin, LRRC32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2615 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14392 | |
LRRC32 | |
LRRC32 Fusion Protein Ag23423 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |