missing translation for 'onlineSavingsMsg'
Learn More

LRRC32 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26021-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16804555

  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.
Specifications

Specifications

LRRC32
Polyclonal
Unconjugated
LRRC32
D11S833E, GARP, Garpin, LRRC32
Rabbit
Antigen Affinity Chromatography
RUO
2615
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14392
LRRC32
LRRC32 Fusion Protein Ag23423
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers