missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRG1 Recombinant Rabbit Monoclonal Antibody (JB34-31)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534656
This item is not returnable.
View return policy
Description
Positive Control: Human serum, human kidney tissue.
The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation.
Specifications
| LRG1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P02750 | |
| LRG1 | |
| Synthetic peptide within Human LRG1 aa 261-310. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| JB34-31 | |
| Unconjugated | |
| LRG1 | |
| 1300008B03Rik; 2310031E04Rik; A2GL; HMFT1766; leucine rich alpha 2 glycoprotein; leucine rich alpha-2-glycoprotein 1; leucine-rich alpha-2-glycoprotein; leucine-rich alpha-2-glycoprotein 1; Leucine-rich alpha-2-glycoprotein precursor; LRG; LRG 1; LRG1; LRG-1; Lrhg | |
| Rabbit | |
| Protein A | |
| RUO | |
| 116844 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction