Learn More
IKA™ LR 1000.3 Laboratory Reactor Vessel
Product Code. 15879871 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15879871 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15879871 Supplier IKA™ Supplier No. 0025001955
Estimated Shipment: 11-12-2026
Ideally suited for optimizing and reproducing chemical reactions, mixing, dispersing, and homogenization processes on a laboratory scale.
  • The ultimate systems when it comes to optimizing and reproducing chemical reaction, mixing, dispersing, and homogenization processes on a laboratory scale.
  • Can be individually customized to perform numerous tasks, including: production of creams and lotions, and the grinding and disintegrating of solids and fibers in liquids or polymers.
  • Single-walled or double-walled vessels made from borosilicate glass or stainless steel are available, each with or without bottom discharge valve.
  • Functionality, safety, and longevity are the main goals in the development of IKA laboratory reactors.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Material FFKM, PEEK, Borosilicate Glass (3.3)
Includes 6 Connectors
Product Type Reactor vessel
Standard Taper Joint 2 x NS 29, 2 x NS 14, 2 x GL 14