Learn More
Buerkle™ LPDE Wide Mouth Wash Bottles
Shop All Bürkle Products
Click to view available options
Closure Color:
Blue
Green
Orange
Red
Transparent
Yellow
Unit Size:
Each
Description
- Wide mouth opening for safe filling
- Yellow translucent color with light protection
- Curved aperture on screw cap for precision jet dispensing without splatters
- Color coded closures
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Translucent Yellow |
| Diameter (Metric) Inner Neck | 39 mm |
| Material | LDPE |
| Closure Color | Green |
| Closure Included | Yes |
| Mouth | Wide |
| Product Type | Wash Bottle |
| Chemical Label | Methanol |