Learn More
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles
Shop All Bürkle Products
Click to view available options
Length (Metric):
10 m
100 m
25 m
50 m
Material:
Copper
Polypropylene
Stainless Steel
Stainless Steel (V2A)
Diameter (Metric) Outer:
1.25 mm
2 mm
2.3 mm
4.5 mm
Unit Size:
Each
Description
- Good chemical resistance
- For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
- Tensile load max. up to 10kg
Specifications
Specifications
| Product Type | Lowering Cable |
| Material | Stainless Steel (V2A) |
| Finish | PTFE-coated |
| For Use With (Application) | Dipping Samplers and Immersion Cylinders, Manually-Operated Drum Reels |
| Diameter (Metric) Outer | 2 mm |
| Length (Metric) | 25 m |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion