Learn More
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles
Product Code. 10498421 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
10 m
100 m
25 m
50 m
Material:
Copper
Polypropylene
Stainless Steel
Stainless Steel (V2A)
Diameter (Metric) Outer:
1.25 mm
2 mm
2.3 mm
4.5 mm
Unit Size:
Each
Product Code. Length (Metric) Material Diameter (Metric) Outer unitSize
10498421 25 m Stainless Steel (V2A) 1.25 mm Each
11882663 10 m Copper 4.5 mm Each
11892663 25 m Copper 4.5 mm Each
11802673 50 m Copper 4.5 mm Each
10066361 50 m Polypropylene 2 mm Each
10157871 25 m Polypropylene 2 mm Each
10279771 10 m Polypropylene 2 mm Each
10107781 10 m Stainless Steel (V2A) 1.25 mm Each
10084800 10 m Stainless Steel (V2A) 2 mm Each
10779972 25 m Stainless Steel (V2A) 2 mm Each
10408431 50 m Stainless Steel (V2A) 1.25 mm Each
10381132 50 m Stainless Steel (V2A) 2 mm Each
17270574 25 m - - Each
10579902 10 m Stainless Steel 2.3 mm Each
17280574 100 m - - Each
15 options
Product Code. 10498421
Brand: Buerkle™ 53090302
Only null left
Add to basket
Add to basket
Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.
  • Good chemical resistance
  • For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
  • Tensile load max. up to 10kg
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lowering Cable
Material Stainless Steel (V2A)
For Use With (Application) Dipping Samplers and Immersion Cylinders, Manually-Operated Drum Reels
Diameter (Metric) Outer 1.25 mm
Length (Metric) 25 m