Learn More
Thermo Scientific LOWENSTEIN KIT 150MM TUBES
Brand:  Thermo Scientific 50128265
LOWENSTEIN KIT 150MM TUBES
Product Code. 11839143
  • ONLINE EXCLUSIVE PRICE
    2685.00 NOK
    2515.24 NOK / Each
    Save 169.76 NOK
    6% Off