Learn More
Low Temperature Viscosity Standard, Paragon Scientific™
Click to view available options
Viscosity:
151216.0 mPa.s (-40°C)
172195 mPa.s (-13°C)
5989.0 mPa.s (-40°C)
757.0 mPa.s (-40°C)
Unit Size:
500mL
Description
Specifications
Specifications
| Chemical Name or Material | N1400B |
| Viscosity | 172195 mPa.s (-13°C) |
| Certifications/Compliance | ISO 17025, UKAS ISO 17034 |
| Quantity | 500 mL |
Nominal values quoted