missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Low Input RiboMinus™ Eukaryote System v2

Product Code. 15800843
Change view
Click to view available options
Quantity:
12 reactions
Unit Size:
12 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15800843 12 reactions 12 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15800843 Supplier Invitrogen™ Supplier No. A15027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Low Input RiboMinus™ Eukaryote System v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA.

Features of the system include:
• Magnetic bead-based purification for rapid, seamless purification
• 1-hour workflow
• Low input range of 100 ng–1 μg

The Low Input RiboMinus™ Eukaryote System v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA. When used with our library construction kits for sequencing, the Low Input RiboMinus™ Eukaryote System v2 provides maximum mapping to reference database with minimum bias, maintaining relative gene expression levels of even smaller RNA species. The resulting total RNA after depletion contains minimum contaminating rRNA with reduced variability.

The Low Input RiboMinus Eukaryote System v2 is designed for the removal of human, mouse, and rat rRNA. A magnetic bead-based purification module is included for purification of the whole transcriptome without altering the expression levels of small or non-coding RNA. For column-based purification, please see the RiboMinus™ Eukaryote Kit v2. The Low Input RiboMinus Eukaryote System v2 has an RNA input range of 100 ng–1 μg. For high inputs (up to 5 ug), the RiboMinus™ Eukaryote System v2 is recommended.

The Low Input RiboMinus™ Eukaryote System v2 contains sufficient reagents to perform 12 ribosomal RNA depletions.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • RiboMinus™ Eukaryote Probe Mix, 2-20°C
• RiboMinus™ Magnetic Beads, 4°C
• 2 x Hybridization Buffer, room temperature
• Binding Solution Concentrate: room temperature
• Wash Solution Concentrate, room temperature
• Nucleic Acid Binding Beads: 4°C
Species Human, Mouse, Rat
Final Product Type Transcriptome RNA
For Use With (Equipment) Ion PGM™ System, Ion Proton™ System, Illumina™ Genome Analyzer II (GA II)
No. of Reactions 12 reactions
Product Line RiboMinus
Quantity 12 reactions
Sequencing Type Transcriptome Sequencing
Starting Material Amount 100 ng - 1 μg total RNA
Workflow Step Sample Enrichment

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.