Learn More
Lenz Laborglasinstrumente™ Low-Form Weighing Bottles
Description
- With interchangeable NS stopper or cap
- With fused-in size (mm) and capacity (mL)
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 45 mL |
| Form | Low |
| No. per Pack | 1 Pack |
| Height (Metric) | 30 mm |
| Diameter (Metric) | 60 mm |
| Closure Type | Button Cover |
| Product Type | Weighing Bottle |