Learn More
Thermo Scientific Chemicals Low Form Nickel Crucible;Cap (ml), 35;Top Dia (mm), 46;Bottom Dia (mm), 38;Depth (mm), 30
Product Code. 15413635
Change view
Click to view available options
Capacity (Metric):
15 mL
20 mL
25 mL
35 mL
55 mL
100 mL
250 mL
Diameter (Metric) Outer:
33mm
36mm
45mm
46mm
47mm
59mm
82mm
Height (Metric):
23mm
30mm
35mm
43mm
46mm
60mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Height (Metric) unitSize
15413635 100 mL 59mm 46mm Each
15413585 15 mL 36mm 23mm Each
15483585 20 mL 33mm 30mm Each
15443595 25 mL 45mm 23mm Each
15403605 35 mL 46mm 30mm Each
15463605 55 mL 46mm 35mm Each
15423615 55 mL 47mm 43mm Each
15443635 250 mL 82mm 60mm Each
8 options
Product Code. 15413635
Brand: Thermo Scientific Chemicals 036057.KT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Low Form
Material Nickel
Capacity (Metric) 100 mL
Diameter (Metric) Outer 59mm
Height (Metric) 46mm
Product Type Nickel Crucible
Diameter (Metric) Base 50mm