Description
- Manufactured from soda lime glass.
- Includes polypropylene screw cap with white elastomer seal and GL thread.
- Autoclavable (121°C).
Specifications
Specifications
| Closure Material | Polypropylene |
| Length (Metric) | 100 mm |
| Material | Soda Lime Glass |
| Quantity | 100/Pk. |
| Thread Size | GL 14 |
| Diameter (Metric) | 12 mm |
| Closure Type | Screw Cap |
| Product Type | Culture Tube |