Learn More
Kartell™ Low Form Beaker with Blue Graduations
Product Code. p-8005802
Change view
Click to view available options
Capacity (Metric):
25 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 16
Pack of 2
Pack of 20
Pack of 4
Product Code. Capacity (Metric) unitSize
10654432 25 mL Pack of 20
10555811 100 mL Pack of 12
10041071 250 mL Pack of 16
10431765 500 mL Each
10063734 1000 mL Each
10691742 2000 mL Pack of 4
10601752 3000 mL Pack of 2
10070941 5000 mL Pack of 2
8 options
Product Code. 10654432
Brand: Kartell™ 0172000
Only null left
Add to basket
Add to basket
Accurately measures the volume of a liquid. Kartell™ Low Form Beaker with Blue Graduations uses horizontal marked lines to assist in measuring out various volumes of a liquid.
  • Good chemical resistance
  • Permanent blue graduations moulded into beaker
  • Can withstand temperatures up to 170°C for short periods
  • Suitable for applications with food
Specifications
Color Crystal Clear
Diameter (Metric) Outer 34mm
Capacity (Metric) 25 mL
Material PMP
Graduation Range 1mL
Height (Metric) 49mm
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Certifications/Compliance ISO 7056-1981(E), BS 5404 Part 1
Scale Permanent Graduated Scale